missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Polypropylene Luer Fitting

Featuring a variety of fitting options, these Luer fittings are ideal for many applications including syringes or small tubing. Cole-Parmer™ Polypropylene Luer Fitting are available with standard, wide, and low-profile barbs.

30.55 € - 61.20 €

Specifications

Certifications/Compliance ISO 594-1 standards
Material Polypropylene
Packaging Quantity 25 Pack
Product Type Luer Fitting
View More Specs

Products 6
Product Code Brand For Use With Price Quantity & Availability  
Product Code Brand For Use With Price Quantity & Availability  
15114865
View Documents
Masterflex™
45508-66
Female luer x 1/4 to 28 in. UNF thread This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15305641
View Documents
Masterflex™
45508-26
Female Luer to Female Luer, elbow
40.90 €
Pack of 25
Estimated Shipment: 21-03-2025
to see stock.
Add to basket
 
15116116
View Documents
Masterflex™
45508-56
Female x Female x Female
39.00 €
Pack of 25
Estimated Shipment: 09-07-2025
to see stock.
Add to basket
 
15102954
View Documents
Masterflex™
45518-82
Male Luer lock to 1/4 to 28 UNF thread
30.70 €
Pack of 25
Estimated Shipment: 21-03-2025
to see stock.
Add to basket
 
15395631
View Documents
Masterflex™
45518-84
Male Luer lock to 10 to 32 thread
30.55 €
Pack of 25
Estimated Shipment: 21-03-2025
to see stock.
Add to basket
 
15329612
View Documents
Masterflex™
45508-85
Male x Female x Female
61.20 €
Pack of 25
Estimated Shipment: 09-07-2025
to see stock.
Add to basket
 
Description

Description

  • Three barb styles: standard, wide-bore, and low-profile.
    • Wide-bore barbs offer enhanced fluid flow through the fitting and are usable with any flexible tubing.
    • Low-profile barbs are ideal for use with semi-rigid tubing, such as nylon, polyethylene, or polypropylene.
  • Manufactured in compliance with ISO 594-1 standards, these fittings are cleaned, inspected and fully traceable.
  • All parts meet tight dimensional tolerances for increased reliability and process purity.
  • Choose from a range of sizes, 1/16in. to 1/4in., with a variety of connection options.
  • Pack of 25
Specifications

Specifications

ISO 594-1 standards
Polypropylene
25 Pack
Luer Fitting
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.