missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polypropylene Microplates, 384-well
GREENER_CHOICE

Offers higher well density for economical use of expensive reagents. Fisherbrand™ Polypropylene Microplates, 384-well include round-shaped wells for optimal sample recovery.

Brand:  Fisherbrand™ 236300

 View more versions of this product

Product Code. 11987954

  • 526.00 € / Pack of 120
Estimated Shipment: 19-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Ideal for storage of compounds and nucleic acid libraries
  • Chemical resistant
  • Withstands temperatures from -80° to +80°C
  • Conform to ANSI™ standard footprint for optimal use in automated handling and liquid-dispensing systems
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ANSI™ standard footprint
16 x 24
Polypropylene
384
120/Pk.
Standard
Blue
120 μL
Round
-80°C to +80°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers