Learn More
Eppendorf™ PCR Tubes - Singles and Strips
GREENER_CHOICE
Eppendorf PCR Tubes ensure uniform heat distribution to the sample for optimal PCR performance. The user-friendly design allows for easy opening and provides a secure seal to prevent evaporation.
Brand:  Eppendorf™ 0030124910
254.60 EUR valid until 2024-12-31
Use promo code "21977" to get your promotional price.
 View more versions of this product
Product Code. 15993982
Quantity Price
1 268.00 € / Pack of 120
5 247.55 € / Pack of 120
Explore more special offers
Description
Description
Ensure efficient and homogenous heat transfer with these thin-walled polypropylene tubes. Eppendorf™ Polypropylene PCR Tubes are easy to open, but provide tight sealing to prevent evaporation in PCR.
  • Design ensures an airtight seal to prevent contamination and evaboration.
  • Thin polypropylene walls ensure efficient and homogeneous heat transfer or fast and high-quality PCR results.
  • 8 reaction tubes in strip-format – ideal for small sample volumes. Easily adaptable for automation.
  • Strips are sealable using flat or domed strip-lids.
  • PCR Clean: Certified free from human DNA, DNase, RNase and PCR inhibitors).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.1 mL
Polypropylene
Flat Strip Cap
Yes
PCR Tube Strip
Non-sterile
PCR Clean (Non-sterile)
Autoclavable
Fast PCR Tube Strips