Learn More
Eppendorf™ twin.tec™ PCR Plates
GREENER_CHOICE
Use these plates for automation and high throughput processes as well as quantitative real-time PCR. Eppendorf™ twin.tec™ PCR Plates combine extremely thin walled polypropylene wells for optimal heat transfer and extremely robust polycarbonate frames for rigidity and torque-resistance.
Brand:  Eppendorf™ 0030128680
150.99 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
 View more versions of this product
Product Code. 10100803
  • 159.64 € / Pack of 25
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Use these plates for automation and high throughput processes as well as quantitative real-time PCR. Eppendorf™ twin.tec™ PCR Plates combine extremely thin walled polypropylene wells for optimal heat transfer and extremely robust polycarbonate frames for rigidity and torque-resistance.
  • Raised well rims for effective sealing, also reduces risk of cross-contamination
  • Exceptionally solid and torque-resistant polycarbonate frame
  • Improved well-to-well tolerance
  • Cut-away corner and alphanumeric grid-referencing
  • Certified free of any detectable human DNA, DNase, RNase and PCR inhibitors
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
Autoclavable
Single-use
Skirted
96
150 μL
Yes
25/Pk.
8 x 12
Red
Real time PCR
Polypropylene, Polycarbonate
Sterile
Conical Bottom (V-well)
Ideal for quantitative real time PCR and fit most thermal cyclers
Yes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM211617K Certificate TypeCertificate of Analysis Date16/08/2023 Catalog Number0030128680
Special Offers
Special Offers
Product Content Correction
Product Title
Eppendorf™ twin.tec™ PCR Plates > Red; Skirted