missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Polypropylene Powder Funnels
Constructed of durable, lightweight polypropylene. Brand™ Polypropylene Powder Funnels are an excellent option for the transfer of samples without flow rate restrictions.
Brand: BRAND™ 148040
This item is not returnable.
View return policy
Description
- With short, wide stem
- Provides excellent resistance against thermal shock

Specifications
150 mm | |
Polypropylene | |
Autoclavable | |
Powder funnel |
31 mm | |
42 mm stem | |
Translucent |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BRAND™ Polypropylene Powder Funnels > Diameter (Metric)Top: 150 mm, Height (Metric): 42 mm
Spot an opportunity for improvement?Share a Content Correction