missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Polypropylene Prinzing Ball Valve Lockout

The two halves of the Polypropylene Prinzing Ball Valve Lockout encompass the ball valve lever to secure it from inadvertent activation of the valve.

Brand:  Brady™ 805851

 View more versions of this product

Product Code. 11776785

  • 111.00 € / Each
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The two halves of the device encompass the ball valve lever to secure it from inadvertent activation of the valve.
  • Locks out pipes from 12.70 to 31.75 mm in open and 12.70 to 63.50 mm closed position.
Specifications

Specifications

Valve Lockout
Green
OSHA 1910.147
118.11 x 304.80 x 219.96 mm
Lockout Tagout
219.96 mm
12 in.
BS08A-GN
Polypropylene
Prinzing Ball Valve Lockout
305 x 220 mm
8.66 in.
1 Ea.
304.8 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.