Learn More
Eppendorf™ Polypropylene Protein LoBind Microcentrifuge Tube
GREENER_CHOICE
Store peptide, antibody or virus samples in these LoBind microcentrifuge tubes. Eppendorf™ Polypropylene Protein LoBind Microcentrifuge Tubes are ideal for work in protein research, mass spectrometry and protein array sample preparation.
12.90 € - 20.48 €
Specifications
Closure Material Polypropylene
Material Polypropylene
For Use With (Application) Protein LoBind consumables are best suited for work in protein research, mass spectrometry, protein array sample preparation, and storage of peptide, antibody or virus samples
No. per Pack 100 Tubes (2 bags of 50)
Sterility PCR Clean
View More Specs
Products 3
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10316752
GREENER_CHOICE
View Documents
Eppendorf™
0030108094
0.5 mL
17.12 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
10708704
GREENER_CHOICE
View Documents
Eppendorf™
0030108116
1.5 mL
12.90 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
10718894
GREENER_CHOICE
View Documents
Eppendorf™
0030108132
2 mL
20.48 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
Description
Description
Store peptide, antibody or virus samples in these LoBind microcentrifuge tubes. Eppendorf™ Polypropylene Protein LoBind Microcentrifuge Tubes are ideal for work in protein research, mass spectrometry and protein array sample preparation.
  • Problem free handling with the tiniest volumes/concentrations of a variety of proteins
  • Preliminary handling of tubes by saturation with BSA or siliconisation is not required in order to guarantee the recovery of the proteins in the plastic tube
  • Recovery rates of more than 99% are achievable
  • Enzyme activities, for example, are not reduced by this tube, and denaturing effects caused by interaction with the surface are almost completely excluded
  • No undesired side effects of a surface coating
  • Tubes are available in Eppendorf LoBind™ material for maximum recovery of valuable samples
  • Maximum protein recovery (over 97%, BSA 1μg/mL)
  • Free from surface coating, e.g., silicone
  • Free from DNA, DNase, RNase and ATP (PCR* clean)
  • Special polypropylene material manufactured under optimized processing conditions
  • High degree of centrifugation durability, rated to 18, 000xg
  • High degree of transparency
  • Safe-Lock lid
Specifications
Specifications
Polypropylene
Protein LoBind consumables are best suited for work in protein research, mass spectrometry, protein array sample preparation, and storage of peptide, antibody or virus samples
PCR Clean
Polypropylene
100 Tubes (2 bags of 50)
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Eppendorf™ Polypropylene Protein LoBind Microcentrifuge Tube