missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Polypropylene Protein LoBind Microcentrifuge Tube
GREENER_CHOICE

Store peptide, antibody or virus samples in these LoBind microcentrifuge tubes. Eppendorf™ Polypropylene Protein LoBind Microcentrifuge Tubes are ideal for work in protein research, mass spectrometry and protein array sample preparation.

Brand:  Eppendorf™ 0030108116

12.19 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.


 View more versions of this product

Product Code. 10708704

  • 12.90 € / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Store peptide, antibody or virus samples in these LoBind microcentrifuge tubes. Eppendorf™ Polypropylene Protein LoBind Microcentrifuge Tubes are ideal for work in protein research, mass spectrometry and protein array sample preparation.

  • Problem free handling with the tiniest volumes/concentrations of a variety of proteins
  • Preliminary handling of tubes by saturation with BSA or siliconisation is not required in order to guarantee the recovery of the proteins in the plastic tube
  • Recovery rates of more than 99% are achievable
  • Enzyme activities, for example, are not reduced by this tube, and denaturing effects caused by interaction with the surface are almost completely excluded
  • No undesired side effects of a surface coating
  • Tubes are available in Eppendorf LoBind™ material for maximum recovery of valuable samples
  • Maximum protein recovery (over 97%, BSA 1μg/mL)
  • Free from surface coating, e.g., silicone
  • Free from DNA, DNase, RNase and ATP (PCR* clean)
  • Special polypropylene material manufactured under optimized processing conditions
  • High degree of centrifugation durability, rated to 18, 000xg
  • High degree of transparency
  • Safe-Lock lid
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Polypropylene
Transparent
Protein LoBind consumables are best suited for work in protein research, mass spectrometry, protein array sample preparation, and storage of peptide, antibody or virus samples
1.5mL
18,000 x g
1.5 mL
Polypropylene
100 Tubes (2 bags of 50)
PCR Clean
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Eppendorf™ Polypropylene Protein LoBind Microcentrifuge Tube > Capacity: 1.5mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.