missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ Polypropylene Reagent Bottle

Store liquid and powder chemicals securely. VITLAB™ Polypropylene Reagent Bottle is constructed of transparent, durable polypropylene for long-term storage. Choose from neck-size stoppers or maximum grip screw caps, both available in a variety of sizes.

Brand:  VITLAB™ 101794

 View more versions of this product

Product Code. 10443101

This item is not returnable. View return policy

Description

Description

Economical and durable plastic reagent bottles are ideal of storing liquids, powders and paste-like materials. A variety of sizes, seals (stopper or screw caps) and neck sizes are available to meet the storage needs of any lab material.
  • Transparent media bottles ideal for long-term storage of chemicals
  • Reagent bottles featuring screw caps or neck stoppers
    • Stoppers: neck-size with a hexagonal-knob cap and red core
    • Screw caps: high-pitch GL threads for significant carrying power
  • Larger sizes (5 and 10L) available with handles for easier carrying
  • Wider mouth bottles are ideal for easier filling, especially of powders and paste-like materials
  • Constructed of study polypropylene
    • Good chemical and temperature resistance
    • Autoclavable at 121 °C (2 bar) according to DIN EN 285
Specifications

Specifications

500 mL
NS stopper made of PP
10 Bottles
Autoclavable
183 mm
87 mm
Long-term storage of liquids
Polypropylene
45/40
Wide
Reagent Bottle
Stopper with Hexagonal Cap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers