missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VITLAB™ Polypropylene Reagent Bottle
Store liquid and powder chemicals securely. VITLAB™ Polypropylene Reagent Bottle is constructed of transparent, durable polypropylene for long-term storage. Choose from neck-size stoppers or maximum grip screw caps, both available in a variety of sizes.
Specifications
For Use With (Application) | Long-term storage of liquids |
---|---|
Material | Polypropylene |
Autoclavable | Autoclavable |
Product Type | Reagent Bottle |
Product Code | Brand | Capacity (Metric) | Closure Type | Diameter (Metric) | Height (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Type | Diameter (Metric) | Height (Metric) | Price | Quantity & Availability | ||
15712046
|
VITLAB™
101594 |
100 mL | Stopper with Hexagonal Cap | 55 mm | 111 mm |
N/A
|
N/A | ||
15291169
|
VITLAB™
101589 |
100 mL | Screw Cap with GL Thread | 55 mm | 96 mm |
N/A
|
N/A | ||
15722046
|
VITLAB™
101694 |
250 mL | Stopper with Hexagonal Cap | 73 mm | 144 mm |
N/A
|
N/A | ||
15271159
|
VITLAB™
100589 |
500 mL | Screw Cap with GL Thread | 87 mm | 165 mm |
N/A
|
N/A | ||
10443101
|
VITLAB™
101794 |
500 mL | Stopper with Hexagonal Cap | 87 mm | 183 mm |
N/A
|
N/A | ||
15281159
|
VITLAB™
100689 |
1000 mL | Screw Cap with GL Thread | 108 mm | 202 mm |
N/A
|
N/A | ||
15732046
|
VITLAB™
101894 |
1000 mL | Stopper with Hexagonal Cap | 108 mm | 214 mm |
N/A
|
N/A | ||
12895733
|
VITLAB™
102089 |
1000 mL | Screw Cap with GL Thread | 105 mm | 197 mm |
N/A
|
N/A | ||
15211179
|
VITLAB™
101989 |
2000 mL | Screw Cap with GL Thread | 131 mm | 243 mm |
N/A
|
N/A | ||
10641353
|
VITLAB™
100789 |
2000 mL | Screw Cap with GL Thread | 131 mm | 245 mm |
N/A
|
N/A | ||
Description
Economical and durable plastic reagent bottles are ideal of storing liquids, powders and paste-like materials. A variety of sizes, seals (stopper or screw caps) and neck sizes are available to meet the storage needs of any lab material.- Transparent media bottles ideal for long-term storage of chemicals
- Reagent bottles featuring screw caps or neck stoppers
- Stoppers: neck-size with a hexagonal-knob cap and red core
- Screw caps: high-pitch GL threads for significant carrying power
- Larger sizes (5 and 10L) available with handles for easier carrying
- Wider mouth bottles are ideal for easier filling, especially of powders and paste-like materials
- Constructed of study polypropylene
- Good chemical and temperature resistance
- Autoclavable at 121 °C (2 bar) according to DIN EN 285
Specifications
Long-term storage of liquids | |
Polypropylene | |
Autoclavable | |
Reagent Bottle |