missing translation for 'onlineSavingsMsg'
Learn More

Brand™ Screw Caps for Microtubes

Manufactured from polypropylene with silicone seal. BRAND™ Screw Caps for Microtubes are available in a variety of colors. For use at temperatures between –196 to 121°C. Autoclavable.

Brand:  Brand™ 780744

130.84 EUR valid until 2024-10-31
Use promo code "21661" to get your promotional price.


 View more versions of this product

Product Code. 10603761

  • 158.93 € / Pack of 1000
Estimated Shipment: 15-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Manufactured from polypropylene with silicone seal.
  • Available in a variety of colors.
  • Temperature range: –196 to 121°C.
  • Autoclavable;
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Screw Cap with Silicone Seal
Polypropylene
Bulk Screw Cap Microtubes 0.5 to 2 mL
Autoclavable
Yellow
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers