Learn More
Cole-Parmer™ Polypropylene Disposable Spatulas, Opaque
Reduce cross-contamination with these polypropylene disposable spatulas. Polypropylene Disposable Spatulas are ideal for chemical and biological material transfer, collection, and manipulation. Spatulas feature both a scoop end and a spatula end.
Brand:  Cole-Parmer™ 06265-12
52.82 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
 View more versions of this product
Product Code. 11989698
  • 55.60 € / Pack of 300
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Reduce cross-contamination with these polypropylene disposable spatulas. Polypropylene Disposable Spatulas are ideal for chemical and biological material transfer, collection, and manipulation. Spatulas feature both a scoop end and a spatula end.
  • Disposable polypropylene (PP) spatulas
  • Use as a spatula, stir rod, or policeman
  • Contoured end is shatter resistant even when exposed to liquid nitrogen
  • Flexible enough to fit into many small containers to scrape or collect samples
  • Hollow core reduces heat and cold transfer
  • Can be used with dilute acids and bases
  • Autoclavable
Specifications
Specifications
300 Pk.
Opaque
Spatula
Polypropylene
Autoclavable
Single-use
For chemical and biological material transfer, collection, and manipulation, used with dilute acids and bases, reduce cross-contamination
222 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer™ Polypropylene Disposable Spatulas, Opaque > Length: 210mm; Opaque