Learn More
Vitlab™ Polypropylene Spray Bottles
Use these bottles for spraying detergents or disinfectants, especially into difficultly accessible areas, as well as applications in thin layer chromatography. VITLAB™ Polypropylene Spray Bottles feature a sprayer insert with stable, smoothly operated pump trigger and adjustable spray nozzle.
Brand: Vitlab™ 53610
Beschreibung
- White polypropylene spray bottles
- Spray nozzle can be regulated from the finest mist (nebulizing) to a precise liquid jet
- Range: approx. 3 to 4 meters
Spezifikation
850 mL | |
Ideal for spraying detergents or disinfectants, especially into difficultly accessible areas, as well as applications in thin layer chromatography | |
5 Pack | |
Yes | |
Spray Bottle | |
Screw Cap |
White | |
Polypropylene | |
White | |
Not Autoclavable | |
Polypropylene |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For research purposes only