Learn More
Kartell™ Polypropylene Stirring Paddles
Constructed of polypropylene and steel for strength, durability and chemical resistance. Kartel™ Polypropylene Stirring Paddles provide a sturdy variety of paddles to fit any stirring need.
Brand:  Kartell™ 0043100
 View more versions of this product
Product Code. 10566961
  • 125.00 € / Pack of 5
Estimated Shipment: 23-10-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Steel rod covered with polypropylene provides a strong connection to the stirrer
  • Steel rods: 8 dia. x 350mmL fits standard overhead stirrer
  • Polypropylene blades are premanently fixed to the rods for stability
    • Fixed propeller for vortex stirring
    • U shape for slower but more vigorous agitation
    • Two sizes of swivel impellers enable the blades to be introduced through a small aperture then fully extend for stirring
Specifications
Specifications
Collapsible paddle, fully extends during stirring
Stirring Paddle
Polypropylene Paddle, Steel Rod Covered with Polypropylene
57 mm W
57 mm