missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Polypropylene Storage Boxes with lid, Autoclavable, Alphanumeric coded
31.61 EUR valid until 2025-05-09
Use promo code "24143" to get your promotional price.
Description
- For 1.5mL, 1.8mL, 2mL vials or 2mL shell vials
- Includes cover
- Dimension (L x W x H): 130 x 130 x 45mm
- 81 cavities with alphanumeric coding of all 4 margins as well as the cavities at the bottom
- Feature alphanumeric indexing for easier vial identification
- Stackable for efficient storage

Specifications
Pink |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Polypropylene Storage Boxes with lid, Autoclavable, Alphanumeric coded > Pink
Spot an opportunity for improvement?Share a Content Correction