missing translation for 'onlineSavingsMsg'
Learn More

Azpack™ Polypropylene Storage Container

Keep food safe and ingredients fresh. Azpack™ Polypropylene Storage Container includes a snap on, airtight lid.

Brand:  Azpack™ 11368053

 View more versions of this product

Product Code. 11368053

  • 7.35 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy

Description

Description

  • Professional food storage quality
  • Lid styled for easy opening and resealing
  • 100% airtight
  • Clear polypropylene material
  • Safe for dishwasher, microwave and fridge/freezer
Specifications

Specifications

370 mL
Snap-on Lid
Polypropylene
Storage
Storage Box
85 x 100 mm (W x H)
95 mm
100 mm
2002/72/EC, BSEN 1186:2002
Clear
Round
Snap-On Lid
1 Each
80 mm
65 to 75 mm
95 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.