missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Plastilab™ Polypropylene Universal Test Tube Racks
BIOPHARMA

Special design allows samples to be incubated at same temperature.

207.00 € - 289.00 €

Specifications

Product Type Test Tube Rack
Autoclavable Autoclavable
Material Polypropylene
For Use With (Application) It can be used dry, wet, in water baths
View More Specs

Products 9
Product Code Brand Depth (Metric) Holds Color Positions Price Quantity & Availability  
Product Code Brand Depth (Metric) Holds Color Positions Price Quantity & Availability  
15855527
BIOPHARMA
View Documents
Kartell™
0056406
246 mm Tubes (13 mm) Yellow 90
214.00 €
Pack of 20
Estimated Shipment: 16-01-2025
to see stock.
Add to basket
 
15865527
BIOPHARMA
View Documents
Kartell™
0056410
246 mm Tubes (13 mm) Red 90
221.00 €
Pack of 20
Estimated Shipment: 13-01-2025
to see stock.
Add to basket
 
15885527
View Documents
Kartell™
0056506
246 mm Tubes (16 mm) Yellow 60
229.00 €
Pack of 20
In Stock
to see stock.
Edge
Add to basket
 
15907860
View Documents
Kartell™
0056804
300 mm Tubes (30 mm) Blue 24
270.00 €
Pack of 20
Estimated Shipment: 13-01-2025
to see stock.
Add to basket
 
15875527
View Documents
Kartell™
0056504
246 mm Tubes (13 mm) Blue 60
207.00 €
Pack of 20
Estimated Shipment: 13-01-2025
to see stock.
Add to basket
 
15895527
View Documents
Kartell™
0056510
246 mm Tubes (16 mm) Red 60
215.00 €
Pack of 20
Estimated Shipment: 13-01-2025
to see stock.
Add to basket
 
15825537
View Documents
Kartell™
0056704
295 mm Tubes (25 mm) Blue 40
250.00 €
Pack of 20
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
15835537
View Documents
Kartell™
0056706
295 mm Tubes (25 mm) Yellow 40
268.00 €
Pack of 20
Estimated Shipment: 29-01-2025
to see stock.
Add to basket
 
15845537
View Documents
Kartell™
0056710
295 mm Tubes (25 mm) Red 40
289.00 €
Pack of 20
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
 
Description

Description

Special design allows samples to be incubated at same temperature.

  • Autoclavable at 121°C
  • Large capacity
  • Can be used dry or wet in waterbaths
  • Does not rot, rust, or deform
  • Alphanumerical grid reference on top tier for easy identification
  • Color-coded for bath work
Specifications

Specifications

Test Tube Rack
Autoclavable
Polypropylene
It can be used dry, wet, in water baths
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.