missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 85, Thermo Scientific Chemicals

14.47 € - 168.84 €

Chemical Identifiers

CAS 9005-70-3
Molecular Formula (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC42H76O7
Molecular Weight (g/mol) NaN
MDL Number MFCD01779641
InChI Key ZGEQUSWSYZZNAI-CLFAGFIQNA-N
Synonym Polyoxyethylene (20) Sorbitan Trioleate, Polysorbate 85
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCOC(=O)CCCCCCC\C=C/CCCCCCCC)C1OCC(OCCO)C1OCCO
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10746432
missing translation for 'viewDocuments'
Thermo Scientific Acros
334150250
25 mL Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10505871
missing translation for 'viewDocuments'
Thermo Scientific Acros
334152500
250 mL Glass bottle
37.89 €
250mL
Please to purchase this item. Need a web account? Register with us today!
10555481
missing translation for 'viewDocuments'
Thermo Scientific Acros
334150010
1 L Glass bottle
103.52 €
1 L
Please to purchase this item. Need a web account? Register with us today!
11966941
missing translation for 'viewDocuments'
Thermo Scientific Acros
334150025
2.5 L Glass bottle
168.84 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

9005-70-3
NaN
ZGEQUSWSYZZNAI-CLFAGFIQNA-N
CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCOC(=O)CCCCCCC\C=C/CCCCCCCC)C1OCC(OCCO)C1OCCO
(C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC42H76O7
MFCD01779641
Polyoxyethylene (20) Sorbitan Trioleate, Polysorbate 85

Specifications

Colorless
9005-70-3
1.0200g/mL
38 to 52mg KOH/g
1.4700 to 1.4720
1.02
300 mPa.s (25°C)
MFCD01779641
Solubility in water: soluble. Other solubilities: 100g/L isopropyl alcohol, xylene and mineral oil, insoluble in propylene glycol
CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCOC(=O)CCCCCCC\C=C/CCCCCCCC)C1OCC(OCCO)C1OCCO
Polysorbate 85
2mg KOH/g max.
Liquid
>200°C
Authentic
83 to 98mg KOH/g
(C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC42H76O7
3% Max.
Polyoxyethylene (20) Sorbitan Trioleate, Polysorbate 85
ZGEQUSWSYZZNAI-CLFAGFIQNA-N
NaN
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Tween]TS 85

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.