missing translation for 'onlineSavingsMsg'
Learn More

Socorex™ Polystyrene Multichannel Round Bottom Reservoirs

Microplate format enables use with automated or robotic pipetting devices.

Brand:  Socorex™ 330.08XL

 View more versions of this product

Product Code. 15560754

  • 130.00 € / Pack of 100
Estimated Shipment: 05-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Polystyrene construction
  • Each channel has a rounded bottom
  • Total capacity 48mL
  • Good bench stability
  • Perfect combination with multichannel pipettes
Specifications

Specifications

48 mL
Non-sterile
8
Polystyrene
Multichannel Reservoir
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers