missing translation for 'onlineSavingsMsg'
Learn More

3M™ E-A-R™ Polyurethane Earplug

Offers high attenuation and are made from a slow expanding, PU foam material. 3M E-A-R Polyurethane Earplug – 36dB.

Brand:  3M™ 7100109118

 View more versions of this product

Product Code. 17227650

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Reduces noise levels by 25dB
  • Fully tested and approved against EN 352-2
  • Made from a soft durable TPE
  • Triple flange design for optimum fit for most ear canal sizes
  • Reusable and no roll down required
  • Finger grip design makes them easy to fit and remove
  • Comes with a sturdy storage case featuring a belt clip
  • 3M™ Earplugs 1271 available with an attachment cord, convenient if you move in and out of noisy environments frequently

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Orange
50/Pk.
Corded
Polyurethane
CE Approved
25 dB
Earplug
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.