Learn More
LABPLAS Polyurethane Sani-Sponge with Buffered Peptone Water
Synthetic polyurethane sponge for sampling surfaces, with buffer to neutralize disinfectant.
Brand: LABPLAS KSS-67815-BPW
Description
- Polyurethane Sani-Sponge
- Saturated with buffered peptone water
- Dimensions: 160 x 229 mm
- Holds: 600 mL
- Option to include gloves with kit
Specifications
Hygiene Sponge Sampling Kit | |
Neutralizers | |
Sterile | |
Gloves |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.