missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LABPLAS Polyurethane Sani-Sponge with Buffered Peptone Water
Synthetic polyurethane sponge for sampling surfaces, with buffer to neutralize disinfectant.
258.00 € - 301.00 €
Specifications
Product Type | Hygiene Sponge Sampling Kit |
---|---|
Media Type | Neutralizers |
Sterility | Sterile |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
17966749
|
LABPLAS
KSS-67810-BPW |
None |
258.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17966469
|
LABPLAS
KSS-67815-BPW |
Gloves |
301.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Polyurethane Sani-Sponge
- Saturated with buffered peptone water
- Dimensions: 160 x 229 mm
- Holds: 600 mL
- Option to include gloves with kit
Specifications
Hygiene Sponge Sampling Kit | |
Neutralizers | |
Sterile |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
LABPLAS Polyurethane Sani-Sponge with Buffered Peptone Water