missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LABPLAS Polyurethane Sani-Sponge with Neutralizing Buffer
Synthetic polyurethane sponge for sampling surfaces, with buffer to neutralize disinfectant.
Brand: LABPLAS KSS-67815-NE
This item is not returnable.
View return policy
Description
- Polyurethane Sani-Sponge
- Saturated with neutralizing buffer
- Dimensions: 160 x 229 mm
- Holds: 600 mL
- Option to include gloves with kit
Specifications
Hygiene Sponge Sampling Kit | |
Neutralizers | |
Sterile | |
Gloves |