missing translation for 'onlineSavingsMsg'
Learn More

LABPLAS Polyurethane Sani-Sponge with Neutralizing Buffer

Synthetic polyurethane sponge for sampling surfaces, with buffer to neutralize disinfectant.

258.00 € - 301.00 €

Specifications

Product Type Hygiene Sponge Sampling Kit
View More Specs

Products 2
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
17926649
View Documents
LABPLAS
KSS-67810-NE
None
258.00 €
Pack of 100
Please to purchase this item. Need a web account? Register with us today!
17936549
View Documents
LABPLAS
KSS-67815-NE
Gloves
301.00 €
Pack of 100
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Polyurethane Sani-Sponge
  • Saturated with neutralizing buffer
  • Dimensions: 160 x 229 mm
  • Holds: 600 mL
  • Option to include gloves with kit
Specifications

Specifications

Hygiene Sponge Sampling Kit
Videos
SDS
Documents

Documents

Product Certifications