Learn More
LABPLAS Polyurethane Sani-Stick with Dey-Engley Neutralizing Broth
Synthetic polyurethane sponge with stick for sampling surfaces, with buffer to neutralize disinfectant.
Brand: LABPLAS KSS-67910-DE
Description
- Polyurethane Sani-Stick
- Saturated with Dey-Engley neutralizing broth
- Dimensions: 160 x 229 mm
- Holds: 600 mL
- Option to include gloves with kit
Specifications
Hygiene Sponge Sampling Kit | |
Neutralizers | |
Sterile | |
None |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.