Learn More
LABPLAS Polyurethane Sani-Stick with Dey-Engley Neutralizing Broth
Synthetic polyurethane sponge with stick for sampling surfaces, with buffer to neutralize disinfectant.
271.00 € - 326.00 €
Specifications
Product Type Hygiene Sponge Sampling Kit
Media Type Neutralizers
Sterility Sterile
View More Specs
Products 2
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
17956549
View Documents
LABPLAS
KSS-67910-DE
None
271.00 €
Pack of 100
17936649
View Documents
LABPLAS
KSS-67915-DE
Gloves
326.00 €
Pack of 100
Description
Description
  • Polyurethane Sani-Stick
  • Saturated with Dey-Engley neutralizing broth
  • Dimensions: 160 x 229 mm
  • Holds: 600 mL
  • Option to include gloves with kit
Specifications
Specifications
Hygiene Sponge Sampling Kit
Neutralizers
Sterile
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
LABPLAS Polyurethane Sani-Stick with Dey-Engley Neutralizing Broth