Learn More
Thermo Scientific™ Ponceau S Staining Solution
Ponceaus S Staining Solution is a ready-to-use membrane stain for evaluating the transfer efficiency of a western blot.
64.50 € - 249.00 €
Specifications
Detection Method | Colorimetric |
---|---|
Formulation | 0.1% (w/v) in 5% acetic acid |
For Use With (Application) | Western Blotting |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
17435103
|
Thermo Scientific™
A40000279 |
Ponceau S Staining Solution, 500 mL |
64.50 €
500mL |
In Stock
Log in to see stock. |
|||||
17641536
|
Thermo Scientific™
A40000278 |
Ponceau S Staining Solution, 1000 mL |
94.75 €
1000mL |
In Stock
Log in to see stock. |
|||||
30088667
|
Thermo Scientific™
A40000278X3 |
Ponceau S Staining Solution, 3 x 1000 mL |
249.00 €
Pack of 3 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ponceaus S Staining Solution is a ready-to-use membrane stain for evaluating the transfer efficiency of a western blot. This stain is compatible with nitrocellulose and PVDF membranes. Ponceau S staining is reversible and can be removed with a short incubation in 0.1% NaOH.
Ponceaus S Staining Solution is a ready-to-use membrane stain for evaluating the transfer efficiency of a western blot. This stain is compatible with nitrocellulose and PVDF membranes. Ponceau S staining is reversible and can be removed with a short incubation in 0.1% NaOH.Features include:
• A nondestructive, reversible method to stain and detect proteins on nitrocellulose and PVDF membranes
• Minimal nonspecific interactions with the membrane surface; provides a reliable method for visualization of protein transfer to a membrane
• Simple and quick staining protocol that results in reddish-pink bands that are easily photographed
• Reversed easily in less than five minutes
• Subsequent western blot detection unaffected as the stain does not alter the membrane-bound protein
Specifications
Colorimetric | |
0.1% (w/v) in 5% acetic acid | |
Western Blotting |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.