Learn More
Haldenwanger™ Porcelain Basins
Use these dishes for a variety of applications. Haldenwanger™ Porcelain Basins feature a smooth internal finish, constructed from optimized laboratory porcelain.
Brand:  Haldenwanger™ 33C/4B
 View more versions of this product
Product Code. 10048160
  • 19.90 € / Each
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Shallow form porcelain basin
  • Available in a variety of sizes
  • Complies with DIN VDE 0335, type C 100
Specifications
Specifications
Porcelain
Evaporating Basin
Smooth
70 mm
Rectangular
Laboratories
70 x 46 x 13 mm
13 mm