Learn More
Haldenwanger™ Porcelain Basins
Use these dishes for a variety of applications. Haldenwanger™ Porcelain Basins feature a smooth internal finish, constructed from optimized laboratory porcelain.
11.80 € - 27.15 €
Specifications
Material Porcelain
Shape Rectangular
Type Evaporating Basin
For Use With (Application) Laboratories
Interior Finish Smooth
View More Specs
Products 4
Product Code Brand Height (Metric) Length (Metric) Price Quantity & Availability  
Product Code Brand Height (Metric) Length (Metric) Price Quantity & Availability  
10509031
View Documents
Haldenwanger™
33C/00
10 mm 45 mm
11.80 €
Each
Estimated Shipment: 09-12-2024
to see stock.
Add to basket
10048160
View Documents
Haldenwanger™
33C/4B
13 mm 70 mm
19.90 €
Each
Estimated Shipment: 12-12-2024
to see stock.
Add to basket
10260961
View Documents
Haldenwanger™
33C/00B
15 mm 50 mm
16.70 €
Each
Estimated Shipment: 19-12-2024
to see stock.
Add to basket
10038150
View Documents
Haldenwanger™
33C/7
16 mm 96 mm
27.15 €
Each
Estimated Shipment: 03-01-2025
to see stock.
Add to basket
Description
Description
  • Shallow form, round-bottom porcelain basin
  • Available in a variety of sizes
  • Includes spout for easy pouring
  • Complies with DIN VDE 0335, type C 100
Specifications
Specifications
Porcelain
Evaporating Basin
Smooth
Rectangular
Laboratories