missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Porcelain Evaporating Dishes

Designed with glazed laboratory porcelain to provide excellent thermal and chemical resistance. Haldenwanger™ Porcelain Evaporating Dishes provide a solid basin for evaporating excessive solvents to produce a concentrated solution or solid precipitate.

Brand:  Haldenwanger™ 109/0

 View more versions of this product

Product Code. 10443001

  • 15.15 € / Each
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Available in shallow and semi-shallow form
  • Sizes up to 175mL glazed; 1,000mL and larger are glazed except outside base
Specifications

Specifications

Round
Porcelain
Evaporating Dish
Glazed
80 mm
75 mL
Round
Laboratories
32 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.