missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Post and Triangular Plastic Base

The Post and Triangular Plastic Base is ideal for controlling and directing foot traffic. Suitable for warehouses, factories, offices etc. Chains and ribbons can be attached to the sturdy 900 mm posts. Includes post and triangular plastic base (no cement).

Brand:  Brady™ 292240

 View more versions of this product

Product Code. 11751316

  • 141.00 € / 1 pack
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Ideal for controlling and directing foot traffic.
  • Suitable for warehouses, factories, offices etc.
  • Chains and ribbons can be attached to the sturdy 900 mm posts.
  • Post and Triangular plastic base (no cement).
Specifications

Specifications

BRADYPOST & NO CEMENT BASE YELL/BLK
35.43 in.
4 Pk.
Black and Yellow Striped
40 mm
900 mm
Triangular Base
Post with Triangular Plastic Base
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.