Learn More
Potassium acrylate hydrate, 95%, Thermo Scientific Chemicals
36.20 € - 146.00 €
Chemical Identifiers
CAS | 10192-85-5 |
---|---|
Molecular Formula | C3H3KO2 |
Molecular Weight (g/mol) | 110.15 |
MDL Number | MFCD00067206 |
InChI Key | ZUBIJGNKOJGGCI-UHFFFAOYSA-M |
Synonym | potassium acrylate, potassium polyacrylate, 2-propenoic acid, potassium salt, potassium prop-2-enoate, 79-10-7 parent, polyacrylic acid, potassium salt, 2-propenoic acid, homopolymer, potassium salt, acrylate potassium, acrylic acid homopolymer potassium salt, acmc-20ajv9 |
PubChem CID | 4429391 |
IUPAC Name | potassium;prop-2-enoate |
SMILES | [K+].[O-]C(=O)C=C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11322427
|
Thermo Scientific Alfa Aesar
L10079.06 |
5 g | N/A | ||||||
11332427
|
Thermo Scientific Alfa Aesar
L10079.14 |
25 g |
146.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10192-85-5 | |
110.15 | |
ZUBIJGNKOJGGCI-UHFFFAOYSA-M | |
4429391 | |
[K+].[O-]C(=O)C=C |
C3H3KO2 | |
MFCD00067206 | |
potassium acrylate, potassium polyacrylate, 2-propenoic acid, potassium salt, potassium prop-2-enoate, 79-10-7 parent, polyacrylic acid, potassium salt, 2-propenoic acid, homopolymer, potassium salt, acrylate potassium, acrylic acid homopolymer potassium salt, acmc-20ajv9 | |
potassium;prop-2-enoate |
Specifications
10192-85-5 | |
C3H3KO2 | |
3695913 | |
ZUBIJGNKOJGGCI-UHFFFAOYSA-M | |
potassium;prop-2-enoate | |
4429391 | |
95% |
>300°C | |
MFCD00067206 | |
potassium acrylate, potassium polyacrylate, 2-propenoic acid, potassium salt, potassium prop-2-enoate, 79-10-7 parent, polyacrylic acid, potassium salt, 2-propenoic acid, homopolymer, potassium salt, acrylate potassium, acrylic acid homopolymer potassium salt, acmc-20ajv9 | |
[K+].[O-]C(=O)C=C | |
110.15 | |
110.16 (Anhydrous) | |
Potassium acrylate hydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Warning
missing translation for 'einecsNumber' : 233-473-9
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only