missing translation for 'onlineSavingsMsg'
Learn More

Potassium Biphthalate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

High quality chemicals for laboratory and specialized industrial use

171.00 € - 326.00 €

Chemical Identifiers

CAS 877-24-7
Molecular Formula C8H5KO4
Molecular Weight (g/mol) 204.222
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
Synonym potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic
PubChem CID 23676735
IUPAC Name potassium;2-carboxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15508074
View Documents
Avantor J.T.Baker
2958-00
100 g Poly Bottle
171.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
15518074
View Documents
Avantor J.T.Baker
2958-01
500 g Poly Bottle
326.00 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications
Show More
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium Biphthalate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.