missing translation for 'onlineSavingsMsg'
Learn More

Potassium bis(trimethylsilyl)amide, 0.7M solution in toluene, +99.4%, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 418238000

430.42 EUR valid until 2024-12-31
Use promo code "21665" to get your promotional price.



 View more versions of this product

Product Code. 10642813

  • 487.43 € / 800mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Potassium bis(trimethylsilyl)amide
0.5M solution in toluene
C6H18KNSi2
MFCD00010330
0.8760g/mL
IUBQJLUDMLPAGT-UHFFFAOYSA-N
potassium;bis(trimethylsilyl)azanide
3251421
0.6 to 0.8 M (Total base)
800 mL
Yellow
40949-94-8, 108-88-3
[(CH3)3Si]2NK
0.6 to 0.8M (Total base)
potassium bis trimethylsilyl amide, khmds, potassium hexamethyldisilazide, hexamethyldisilazane potassium salt, potassium bis trimethylsilyl azanide, potassium bis trimethylsilyl amide 1m sol. in thf, potassiumbis trimethylsilyl amide, hexamethyldislazane potassium salt, 1,1,1,3,3,3-hexamethyldisilazane potassium salt, potassium bis-trimethylsilylamide
C[Si](C)(C)[N-][Si](C)(C)C.[K+]
199.485
199.49
AcroSeal™ Glass Bottle
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Potassium bis(trimethylsilyl)amide, 0.7M (15 wt.%) solution in toluene

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H350-May cause cancer.
  • H341-Suspected of causing genetic defects.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
  • H412-Harmful to aquatic life with long lasting effects.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only