missing translation for 'onlineSavingsMsg'
Learn More

Potassium bis(trimethylsilyl)amide, 0.7M solution in toluene, +99.4%, Thermo Scientific Chemicals

109.55 € - 487.43 €

Specifications

Chemical Name or Material Potassium bis(trimethylsilyl)amide
Color Yellow
Name Note 0.5M solution in toluene
CAS 40949-94-8, 108-88-3
Molecular Formula C6H18KNSi2
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10742623
View Documents Promotion Details
Thermo Scientific Acros
418231000
100 mL AcroSeal™ Glass bottle
109.55 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10642813
View Documents Promotion Details
Thermo Scientific Acros
418238000
800 mL AcroSeal™ Glass Bottle
487.43 €
800mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Potassium bis(trimethylsilyl)amide
0.5M solution in toluene
C6H18KNSi2
MFCD00010330
0.8760g/mL
IUBQJLUDMLPAGT-UHFFFAOYSA-N
potassium;bis(trimethylsilyl)azanide
3251421
0.6 to 0.8 M (Total base)
Yellow
40949-94-8, 108-88-3
[(CH3)3Si]2NK
0.6 to 0.8M (Total base)
potassium bis trimethylsilyl amide, khmds, potassium hexamethyldisilazide, hexamethyldisilazane potassium salt, potassium bis trimethylsilyl azanide, potassium bis trimethylsilyl amide 1m sol. in thf, potassiumbis trimethylsilyl amide, hexamethyldislazane potassium salt, 1,1,1,3,3,3-hexamethyldisilazane potassium salt, potassium bis-trimethylsilylamide
C[Si](C)(C)[N-][Si](C)(C)C.[K+]
199.485
199.49
Liquid
Videos
Safety and Handling

Safety and Handling

GHS P Statement Causes severe skin burns and eye damage. May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure. Suspected of damaging the unborn child. Highly flammable liquid and vapor. Suspected of causing cancer. Suspected of causing genetic defects if inhaled. Harmful to aquatic life with long lasting effects.

GHS P Statement IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. Obtain special instructions before use. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Avoid release to the environment.

Danger

Product Identifier
  • Potassium bis(trimethylsilyl)amide, 0.7M (15 wt.%) solution in toluene

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H350-May cause cancer.
  • H341-Suspected of causing genetic defects.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
  • H412-Harmful to aquatic life with long lasting effects.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only