Learn More
Potassium borohydride, 98+%, Thermo Scientific Chemicals
17.99 € - 1000.00 €
Chemical Identifiers
CAS | 13762-51-1 |
---|---|
Molecular Formula | BH4K |
Molecular Weight (g/mol) | 53.94 |
MDL Number | MFCD00011396 |
InChI Key | ICRGAIPBTSPUEX-UHFFFAOYSA-N |
Synonym | potassium borohydride, potassium tetrahydroborate, potassium borohydrate, unii-0ws230dtgf, potassium tetrahydroborate 1-, borohydrure de potassium french, borate 1-, tetrahydro-, potassium, 0ws230dtgf, borohydrure de potassium, borylpotassium |
PubChem CID | 22892188 |
SMILES | [BH4-].[K+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10311394
|
Thermo Scientific Acros
196550050 |
5 g | Glass bottle |
17.99 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10646592
|
Thermo Scientific Acros
196551000 |
100 g | Glass bottle |
90.85 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10123880
|
Thermo Scientific Acros
196555000 |
500 g | Glass bottle |
319.59 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10647934
|
Thermo Scientific Acros
196550025 |
2.5 kg | Plastic drum |
1000.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentChemical Identifiers
13762-51-1 | |
53.94 | |
ICRGAIPBTSPUEX-UHFFFAOYSA-N | |
22892188 |
BH4K | |
MFCD00011396 | |
potassium borohydride, potassium tetrahydroborate, potassium borohydrate, unii-0ws230dtgf, potassium tetrahydroborate 1-, borohydrure de potassium french, borate 1-, tetrahydro-, potassium, 0ws230dtgf, borohydrure de potassium, borylpotassium | |
[BH4-].[K+] |
Specifications
500.0°C | |
White | |
98+% | |
KBH4 | |
04,398; 06,476; 08,407 | |
potassium borohydride, potassium tetrahydroborate, potassium borohydrate, unii-0ws230dtgf, potassium tetrahydroborate 1-, borohydrure de potassium french, borate 1-, tetrahydro-, potassium, 0ws230dtgf, borohydrure de potassium, borylpotassium | |
ICRGAIPBTSPUEX-UHFFFAOYSA-N | |
potassium;boron(1-) | |
22892188 | |
≥98% |
13762-51-1 | |
Crystalline Powder | |
BH4K | |
MFCD00011396 | |
15, 7737 | |
Solubility in water: 190g/L (25°C). Other solubilities: reacts with water, partially soluble in ammonia, insoluble in ethers, hydrocarbons, isopropylamine,, benzene, hexane, dioxane, thf and acetonitrile, 7g/L methanol (20°C) | |
[BH4-].[K+] | |
53.94 | |
53.93 | |
Potassium borohydride, 98% |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
Causes severe skin burns and eye damage.
Toxic in contact with skin.
In contact with water releases flammable gas.
Reacts violently with water.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 237-360-5
missing translation for 'rtecsNumber' : TS7525000
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.