missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromate-Bromide Solution 0.0167M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™

34.37 € - 78.35 €

Chemical Identifiers

MDL Number 11359
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10479750
SDS View Documents
Fisher Chemical
J/6100/15
1 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10614732
SDS View Documents
Fisher Chemical
J/6100/17
2.5 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

11359

Specifications

Colorless
Liquid
11359
Amber glass bottle
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Potassium bromate-bromide, aqueous solution, 0.0167M (0.1N)

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 1B

Hazard Statement
  • H350-May cause cancer.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P281-Use personal protective equipment as required.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.