Learn More
Potassium bromomethyltrifluoroborate, 95%, Thermo Scientific Chemicals
54.40 € - 180.00 €
Chemical Identifiers
CAS | 888711-44-2 |
---|---|
Molecular Formula | CH2BBrF3K |
Molecular Weight (g/mol) | 200.83 |
InChI Key | AZDFPIRYUOCVCJ-UHFFFAOYSA-N |
Synonym | potassium bromomethyl trifluoroborate, potassium bromomethyltrifluoroborate, potassium bromomethyl trifluoroboranuide, potassium bromomethyl trifluoro boranuide, potassium bromomethyl trifluoro borate 1-, borate 1-, bromomethyl trifluoro-, potassium 1:1 , t-4, pubchem11564, bromomethyl potassium trifluoroborate, postassium bromomethyltrifluoroborate, trifluoro bromomethyl potassioboron v |
PubChem CID | 23690312 |
IUPAC Name | potassium;bromomethyl(trifluoro)boranuide |
SMILES | [B-](CBr)(F)(F)F.[K+] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
888711-44-2 | |
200.83 | |
potassium bromomethyl trifluoroborate, potassium bromomethyltrifluoroborate, potassium bromomethyl trifluoroboranuide, potassium bromomethyl trifluoro boranuide, potassium bromomethyl trifluoro borate 1-, borate 1-, bromomethyl trifluoro-, potassium 1:1 , t-4, pubchem11564, bromomethyl potassium trifluoroborate, postassium bromomethyltrifluoroborate, trifluoro bromomethyl potassioboron v | |
potassium;bromomethyl(trifluoro)boranuide |
CH2BBrF3K | |
AZDFPIRYUOCVCJ-UHFFFAOYSA-N | |
23690312 | |
[B-](CBr)(F)(F)F.[K+] |
Specifications
220.0°C | |
White | |
95% | |
potassium bromomethyl trifluoroborate, potassium bromomethyltrifluoroborate, potassium bromomethyl trifluoroboranuide, potassium bromomethyl trifluoro boranuide, potassium bromomethyl trifluoro borate 1-, borate 1-, bromomethyl trifluoro-, potassium 1:1 , t-4, pubchem11564, bromomethyl potassium trifluoroborate, postassium bromomethyltrifluoroborate, trifluoro bromomethyl potassioboron v | |
[B-](CBr)(F)(F)F.[K+] | |
200.83 | |
200.83 | |
Authentic | |
Potassium bromomethyltrifluoroborate |
888711-44-2 | |
Powder | |
CH2BBrF3K | |
AZDFPIRYUOCVCJ-UHFFFAOYSA-N | |
potassium;bromomethyl(trifluoro)boranuide | |
23690312 | |
95% | |
Glass bottle |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
GHS Signal Word: Warning
- Potassium bromomethyltrifluoroborate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only