Learn More
Potassium carbonate, anhydrous, 99%, Thermo Scientific Chemicals
27.84 € - 221.10 €
Chemical Identifiers
CAS | 584-08-7 |
---|---|
Molecular Formula | CK2O3 |
Molecular Weight (g/mol) | 138.21 |
MDL Number | MFCD00011382 |
InChI Key | BWHMMNNQKKPAPP-UHFFFAOYSA-L |
Synonym | potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate |
PubChem CID | 11430 |
SMILES | [K+].[K+].[O-]C([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11457967
|
Thermo Scientific Alfa Aesar
A16625.36 |
500 g |
27.84 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11467967
|
Thermo Scientific Alfa Aesar
A16625.0E |
2500 g |
71.86 €
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11477967
|
Thermo Scientific Alfa Aesar
A16625.0C |
10,000 g |
221.10 €
10000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potassium carbonate has a wide range of industrial and research applications. The major use of potassium carbonate is the manufacture of chemicals, fertilizers, soap and glass. It is used as a drying agent for solvents especially if there are traces of acids present, and as a scavenger of acids produced in organic synthesis. It is used as a buffering agent during the production of wine. It is also used as a fire suppressant in extinguishers. In organic synthesis, it is an extensively used reagent for the alkylation and arylation reactions on carbon, sulfur, nitrogen, and oxygen centres. It is also used in aldol condensation, Michael reaction, acylation, Wittig reaction, Knoevenagel reaction, elimination reaction, epoxide opening, cyclization, metal-catalyzed cross coupling reaction, hydration, and a wide array of reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPotassium carbonate has a wide range of industrial and research applications. The major use of potassium carbonate is the manufacture of chemicals, fertilizers, soap and glass. It is used as a drying agent for solvents especially if there are traces of acids present, and as a scavenger of acids produced in organic synthesis. It is used as a buffering agent during the production of wine. It is also used as a fire suppressant in extinguishers. In organic synthesis, it is an extensively used reagent for the alkylation and arylation reactions on carbon, sulfur, nitrogen, and oxygen centres. It is also used in aldol condensation, Michael reaction, acylation, Wittig reaction, Knoevenagel reaction, elimination reaction, epoxide opening, cyclization, metal-catalyzed cross coupling reaction, hydration, and a wide array of reactions.
Solubility
Freely soluble in water. Insoluble in acetone, and alcohol.
Notes
Potassium carbonate is a hygroscopic and deliquescent solid.
Chemical Identifiers
584-08-7 | |
138.21 | |
BWHMMNNQKKPAPP-UHFFFAOYSA-L | |
11430 |
CK2O3 | |
MFCD00011382 | |
potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate | |
[K+].[K+].[O-]C([O-])=O |
Specifications
891°C | |
11.5 to 12.5 | |
K2CO3 | |
4267587 | |
potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate | |
BWHMMNNQKKPAPP-UHFFFAOYSA-L | |
dipotassium carbonate | |
11430 | |
99% | |
Hygroscopic | |
Potassium carbonate, anhydrous |
584-08-7 | |
CK2O3 | |
MFCD00011382 | |
14,7619 | |
Freely soluble in water. Insoluble in acetone,and alcohol. | |
[K+].[K+].[O-]C([O-])=O | |
138.21 | |
138.21 | |
Odorless | |
2.29 |
Safety and Handling
GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 209-529-3
missing translation for 'rtecsNumber' : TS7750000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Potassium carbonate anhydrous
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : carbonic acid
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.