missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride, Crystal, Multi-Compendial, U.S.P., J.T. Baker™
130.00 € - 324.00 €
Chemical Identifiers
CAS | 7447-40-7 |
---|---|
Molecular Formula | ClK |
Molecular Weight (g/mol) | 74.55 |
MDL Number | MFCD00011360 |
InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
Synonym | potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs |
PubChem CID | 4873 |
ChEBI | CHEBI:32588 |
SMILES | [Cl-].[K+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15569660
|
Avantor J.T.Baker
3045-01 |
500 g | Poly Bottle |
130.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15568074
|
Avantor J.T.Baker
3045-05 |
2.5 kg | Poly Bottle |
324.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
7447-40-7 | |
74.55 | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
4873 | |
[Cl-].[K+] |
ClK | |
MFCD00011360 | |
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
CHEBI:32588 |
Specifications
7447-40-7 | |
ClK | |
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
[Cl-].[K+] | |
74.55 | |
CHEBI:32588 | |
Multi-Compendial/USP | |
Potassium Chloride |
Crystals | |
MFCD00011360 | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
potassium;chloride | |
4873 | |
74.55 | |
1L = 1.99kg |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 500GR POTASSIUM CHLORIDE,MULTI CGMP Multi-Compendi
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P273-Avoid release to the environment.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Potassium chloride
- UNKNOWN ACUTE-100 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 100% of components with unknown hazards to the aquatic environment.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title