missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride (White Crystals), Fisher BioReagents
31.15 € - 48.60 €
Chemical Identifiers
CAS | 7447-40-7 |
---|---|
Molecular Formula | ClK |
Molecular Weight (g/mol) | 74.55 |
MDL Number | MFCD00011360 |
InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
Synonym | potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs |
PubChem CID | 4873 |
ChEBI | CHEBI:32588 |
IUPAC Name | potassium chloride |
SMILES | [Cl-].[K+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10735874
|
Fisher BioReagents
BP366-500 |
500 g | Poly Bottle |
31.15 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10697623
|
Fisher BioReagents
BP366-1 |
1 kg | Poly Bottle |
48.60 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Potassium Chloride is used in buffers for molecular biology and cell culture applications.Chemical Identifiers
7447-40-7 | |
74.55 | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
4873 | |
potassium chloride |
ClK | |
MFCD00011360 | |
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
CHEBI:32588 | |
[Cl-].[K+] |
Specifications
1420°C | |
7447-40-7 | |
6 | |
99.0 to 100.5 % | |
MFCD00011360 | |
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
[Cl-].[K+] | |
74.55 | |
CHEBI:32588 | |
99.0 to 100.5% | |
99.0-100.5% |
770°C | |
White | |
Solid | |
ClK | |
15, 7742 | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
potassium chloride | |
4873 | |
74.55 | |
Pass Test | |
Potassium Chloride |
Safety and Handling
CAUTION!
Emergency Overview
May cause skin, eye, and respiratory tract irritation. Hygroscopic. Wear personal protective equipment. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.
NFPA
Health:1
Flammability:0
Instability:1
Recommended Storage : RT