missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexacyanocobaltate(III), 95%, Thermo Scientific Chemicals

19.30 € - 387.00 €

Chemical Identifiers

CAS 13963-58-1
Molecular Formula C6CoK3N6
Molecular Weight (g/mol) 332.34
MDL Number MFCD00011391
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10348100
View Documents Promotion Details
Thermo Scientific
203920050
5 g Glass bottle
19.30 €
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10676412
View Documents Promotion Details
Thermo Scientific Acros
203921000
100 g Glass bottle
110.00 €
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10480861
View Documents Promotion Details
Thermo Scientific Acros
203925000
500 g Glass bottle
387.00 €
500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13963-58-1
C6CoK3N6
332.34
MFCD00011391

Specifications

13963-58-1
Crystalline Solid
C6CoK3N6
MFCD00011391
Solubility in water: freely soluble. Other solubilities: insoluble in alcohol
332.34
1.87
Potassium hexacyanocobaltate(III), 95%
Brown to Yellow
95%
K3[Co(CN)6]
15, 7758
332.34
95%
1.8700g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Contact with acids liberates very toxic gas.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 237-742-1

missing translation for 'rtecsNumber' : GF9470000

missing translation for 'tsca' : TSCA

Product Identifier
  • Potassium hexacyanocobaltate(III)

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H373-May cause damage to organs.
  • H302-Harmful if swallowed.
  • EUH032-Contact with acids liberates very toxic gas.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P402+P404-Store in a dry place. Store in a closed container.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only