missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydroxide, ca. 85%, for analysis, pellets, Thermo Scientific Chemicals

22.11 € - 479.39 €

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
SMILES [OH-].[K+]
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10575355
SDS View Documents
Thermo Scientific Acros
134062500
250 g Plastic bottle
22.11 €
250g
Please to purchase this item. Need a web account? Register with us today!
10113190
SDS View Documents
Thermo Scientific Acros
134060010
1 kg Plastic bottle
47.44 €
1kg
Please to purchase this item. Need a web account? Register with us today!
10235140
SDS View Documents
Thermo Scientific Acros
134060025
2.5 kg Plastic bottle
108.54 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
10175723
SDS View Documents
Thermo Scientific Acros
134060100
10 kg Plastic drum
302.00 €
10kg
Please to purchase this item. Need a web account? Register with us today!
10454015
SDS View Documents
Thermo Scientific Acros
134060250
25 kg Fibre drum
479.39 €
25kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium hydroxide, ca. 85%, for analysis, pellets, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.