missing translation for 'onlineSavingsMsg'
Learn More
Potassium hydroxide, Puriss. p.a., Reag. Ph. Eur., ≥85%, pellets, Honeywell Fluka™ Product Code.: 15603070

Potassium hydroxide, Puriss. p.a., Reag. Ph. Eur., ≥85%, pellets, Honeywell Fluka™

Product Code. 15603070 Shop All Honeywell Fluka Products
2.5 kg
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
6 x 500 g
4 x 5 kg
6 x 1 kg
This item is not returnable. View return policy

Product Code. 15603070

Brand: Honeywell-Fluka 306032.5KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a., Reag. Ph. Eur.,≥85%, pellets

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693Show MoreShow Less
PubChem CID 14797
ChEBI CHEBI:32035
SMILES [OH-].[K+]

Specifications

Linear Formula KOH
UN Number 1813
Quantity 2.5 kg
IUPAC Name potassium hydroxide
Formula Weight 56.11g/mol
Percent Purity ≥85%
Grade puriss. p.a.
Vapor Pressure 1mmHg (719°C)
Chemical Name or Material Potassium Hydroxide, Reag. Ph. Eur.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium hydroxide, Puriss. p.a., Reag. Ph. Eur., ≥85%, pellets, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.