missing translation for 'onlineSavingsMsg'
Learn More
Potassium Hydroxide Solution, 0.1M, Fisher Chemical™ Product Code.: 12923584
compliance-icons

Potassium Hydroxide Solution, 0.1M, Fisher Chemical™

Product Code. 12923584
1000 mL
Click to view available options
Quantity:
1000 mL
This item is not returnable. View return policy

Product Code. 12923584

Brand: Fisher Chemical VL 265 1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Concentration 0.1mol/L
Quantity 1000 mL
Physical Form Liquid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1098458 Certificate TypeCertificate of Analysis Date30/03/2025 Catalog NumberVL 265 1000
Lot Number1065187 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberVL 265 1000
Lot Number1040724 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberVL 265 1000
Lot Number1007389 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberVL 265 1000
Lot Number1004846 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog NumberVL 265 1000
compliance-icons
Product Identifier
  • Potassium hydroxide 0.1M (0.1N). Volumetric Solution
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium Hydroxide Solution, 0.1M, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.