missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium iodide, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ≥99.5%, Honeywell Fluka™
Puriss. p.a., Reag. ISO, Reag. Ph. Eur.,≥99.5%
Chemical Identifiers
CAS | 7681-11-0 |
---|---|
Molecular Formula | IK |
Molecular Weight (g/mol) | 166.003 |
MDL Number | MFCD00011405 |
InChI Key | NLKNQRATVPKPDG-UHFFFAOYSA-M |
Synonym | potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide |
PubChem CID | 4875 |
ChEBI | CHEBI:8346 |
IUPAC Name | potassium;iodide |
SMILES | [K+].[I-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15623000
|
Honeywell-Fluka
30315-100G |
100 g |
N/A
|
N/A | |||||
15643000
|
Honeywell-Fluka
30315-250G |
250 g |
N/A
|
N/A | |||||
15663000
|
Honeywell-Fluka
30315-500G |
500 g |
N/A
|
N/A
|
|||||
15693000
|
Honeywell-Fluka
30315-6X100G |
6 x 100 g |
N/A
|
N/A | |||||
15633000
|
Honeywell-Fluka
30315-1KG |
1 kg |
N/A
|
N/A | |||||
15613010
|
Honeywell-Fluka
30315-6X250G |
6 x 250 g |
N/A
|
N/A | |||||
15623010
|
Honeywell-Fluka
30315-6X500G |
6 x 500 g |
N/A
|
N/A | |||||
15683000
|
Honeywell-Fluka
30315-5KG |
5 kg |
N/A
|
N/A | |||||
15603010
|
Honeywell-Fluka
30315-6X1KG |
6 x 1 kg |
N/A
|
N/A | |||||
15653000
|
Honeywell-Fluka
30315-4X5KG |
4 x 5 kg |
N/A
|
N/A | |||||
15673000
|
Honeywell-Fluka
30315-50KG |
50 kg |
N/A
|
N/A | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7681-11-0 | |
166.003 | |
NLKNQRATVPKPDG-UHFFFAOYSA-M | |
4875 | |
potassium;iodide |
IK | |
MFCD00011405 | |
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
CHEBI:8346 | |
[K+].[I-] |
Specifications
7681-11-0 | |
KI | |
NONH for all modes of transport | |
NLKNQRATVPKPDG-UHFFFAOYSA-M | |
potassium;iodide | |
4875 | |
166g/mol | |
ACS/puriss. p.a. | |
Potassium Iodide, Reag. Ph. Eur. reag. ISO |
IK | |
MFCD00011405 | |
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
[K+].[I-] | |
166.003 | |
CHEBI:8346 | |
≥99.5% | |
1mmHg (745°C) |
Safety and Handling
P305 + P351 + P338
H302-H315-H319
missing translation for 'einecsNumber' : 231-659-4
missing translation for 'rtecsNumber' : TT2975000
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- Specific target organ toxicity Category 1
Hazard Statement
- H372-Causes damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.