missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium permanganate solution, Volumetric, 0.1 M KMnO4 (0.5N), Honeywell Fluka™
Volumetric, 0.1 M KMnO4 (0.5N)
163.68 € - 569.28 €
Chemical Identifiers
CAS | 7722-64-7 |
---|---|
Molecular Formula | KMnO4 |
Molecular Weight (g/mol) | 158.032 |
MDL Number | MFCD00011364 |
InChI Key | VZJVWSHVAAUDKD-UHFFFAOYSA-N |
Synonym | potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo |
PubChem CID | 516875 |
IUPAC Name | potassium;permanganate |
SMILES | [O-][Mn](=O)(=O)=O.[K+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15614850
|
Honeywell-Fluka
35201-2.5L |
2.5 L | Glass bottle |
163.68 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15624850
|
Honeywell-Fluka
35201-4X2.5L |
4 x 2.5 L | Glass Bottle |
569.28 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Chemical Identifiers
7722-64-7 | |
158.032 | |
VZJVWSHVAAUDKD-UHFFFAOYSA-N | |
516875 | |
[O-][Mn](=O)(=O)=O.[K+] |
KMnO4 | |
MFCD00011364 | |
potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo | |
potassium;permanganate |
Specifications
7722-64-7 | |
MFCD00011364 | |
potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo | |
[O-][Mn](=O)(=O)=O.[K+] | |
158.032 | |
158.03g/mol |
KMnO4 | |
UN3082 | |
VZJVWSHVAAUDKD-UHFFFAOYSA-N | |
potassium;permanganate | |
516875 | |
Potassium permanganate solution |
Safety and Handling
P273
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Potassium permanganate solution
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : potassium permanganate