missing translation for 'onlineSavingsMsg'
Learn More

Potassium phosphate monohydrate, 95%, Thermo Scientific Chemicals

117.59 € - 398.99 €

Chemical Identifiers

CAS 27176-10-9
Molecular Formula H2K3O5P
Molecular Weight (g/mol) 230.28
MDL Number MFCD00150410
InChI Key RMNIZOOYFMNEJJ-UHFFFAOYSA-K
Synonym tri-potassium phosphate monohydrate, potassium phosphate tribasic monohydrate, potassium phosphate hydrate, potassium phosphate monohydrate, unii-4927m96tfv, tert-potassium phosphate monohydrate, potassium phosphate, tribasic, monohydrate, phosphoric acid, tripotassium salt, monohydrate, tripotassium phosphate, monohydrate, acmc-1chx7
PubChem CID 22031701
IUPAC Name tripotassium;phosphate;hydrate
SMILES O.[O-]P(=O)([O-])[O-].[K+].[K+].[K+]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11358708
View Documents
Thermo Scientific Alfa Aesar
041586.36
500 g
117.59 €
500g
Please to purchase this item. Need a web account? Register with us today!
11368708
View Documents
Thermo Scientific Alfa Aesar
041586.A3
2 kg
398.99 €
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Potassium phosphate monohydrate acts as a reagent and buffer used in molecular biology, biochemistry, and chromatography. It is utilized to study the effects of freezing and thawing of proteins sensitive to conformational changes. It is also used in the extraction of keratohyalin protein from bovine tissue. Further, it is used as a food additive, emulsifier, foaming agent and fertilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

27176-10-9
230.28
RMNIZOOYFMNEJJ-UHFFFAOYSA-K
22031701
O.[O-]P(=O)([O-])[O-].[K+].[K+].[K+]
H2K3O5P
MFCD00150410
tri-potassium phosphate monohydrate, potassium phosphate tribasic monohydrate, potassium phosphate hydrate, potassium phosphate monohydrate, unii-4927m96tfv, tert-potassium phosphate monohydrate, potassium phosphate, tribasic, monohydrate, phosphoric acid, tripotassium salt, monohydrate, tripotassium phosphate, monohydrate, acmc-1chx7
tripotassium;phosphate;hydrate

Specifications

27176-10-9
Granules
MFCD00150410
Soluble in water. Insoluble in alcohol.
O.[O-]P(=O)([O-])[O-].[K+].[K+].[K+]
230.28
230.28 (212.27 Anhydrous)
White granules
Potassium phosphate monohydrate
White
H2K3O5P
tri-potassium phosphate monohydrate, potassium phosphate tribasic monohydrate, potassium phosphate hydrate, potassium phosphate monohydrate, unii-4927m96tfv, tert-potassium phosphate monohydrate, potassium phosphate, tribasic, monohydrate, phosphoric acid, tripotassium salt, monohydrate, tripotassium phosphate, monohydrate, acmc-1chx7
RMNIZOOYFMNEJJ-UHFFFAOYSA-K
tripotassium;phosphate;hydrate
22031701
Odorless
2.564
Videos
Safety and Handling

Safety and Handling

P261-P271-P280-P304+P340-P305+P351+P338-P310-P312-P501c

H318-H335

missing translation for 'einecsNumber' : 231-907-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only