Learn More
Potassium phosphate, tribasic, 97%, pure, anhydrous, Thermo Scientific Chemicals
30.35 € - 423.11 €
Chemical Identifiers
CAS | 7778-53-2 |
---|---|
Molecular Formula | K3O4P |
Molecular Weight (g/mol) | 212.27 |
MDL Number | MFCD00036295 |
InChI Key | LWIHDJKSTIGBAC-UHFFFAOYSA-K |
Synonym | potassium phosphate, tripotassium phosphate, potassium phosphate tribasic, phosphoric acid, tripotassium salt, potassium orthophosphate, tripotassium orthophosphate, caswell no. 700a, potassium phosphate, tribasic, potassium phosphate k3po4, ccris 7321 |
PubChem CID | 62657 |
IUPAC Name | tripotassium;phosphate |
SMILES | [O-]P(=O)([O-])[O-].[K+].[K+].[K+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10463123
|
Thermo Scientific Acros
387680250 |
25 g | Plastic bottle |
30.35 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10573683
|
Thermo Scientific Acros
387685000 |
500 g | Plastic bottle |
58.09 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10349962
|
Thermo Scientific Acros
387680010 |
1 kg | Plastic bottle |
96.08 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10379333
|
Thermo Scientific Acros
387680025 |
2.5 kg | Plastic bottle |
192.96 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13498538
|
Thermo Scientific Acros
387680100 |
10 kg | Plastic drum |
423.11 €
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7778-53-2 | |
212.27 | |
LWIHDJKSTIGBAC-UHFFFAOYSA-K | |
62657 | |
[O-]P(=O)([O-])[O-].[K+].[K+].[K+] |
K3O4P | |
MFCD00036295 | |
potassium phosphate, tripotassium phosphate, potassium phosphate tribasic, phosphoric acid, tripotassium salt, potassium orthophosphate, tripotassium orthophosphate, caswell no. 700a, potassium phosphate, tribasic, potassium phosphate k3po4, ccris 7321 | |
tripotassium;phosphate |
Specifications
1340.0°C | |
White | |
97% | |
K3PO4 | |
15,7781 | |
Solubility in water: 508g/L (25°C). Other solubilities: insoluble in alcohol | |
[O-]P(=O)([O-])[O-].[K+].[K+].[K+] | |
212.27 | |
212.27 | |
Pure |
7778-53-2 | |
Powder, Granules or Chunks | |
K3O4P | |
MFCD00036295 | |
potassium phosphate, tripotassium phosphate, potassium phosphate tribasic, phosphoric acid, tripotassium salt, potassium orthophosphate, tripotassium orthophosphate, caswell no. 700a, potassium phosphate, tribasic, potassium phosphate k3po4, ccris 7321 | |
LWIHDJKSTIGBAC-UHFFFAOYSA-K | |
tripotassium;phosphate | |
62657 | |
97% | |
Potassium phosphate, tribasic, Tribasic 97%, Anhydrous |
Safety and Handling
GHS H Statement
Causes serious eye damage.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-907-1
- Potassium phosphate tribasic
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only