Learn More
Potassium polysulfide, pure, Thermo Scientific Chemicals
42.51 € - 255.27 €
Chemical Identifiers
CAS | 37199-66-9 |
---|---|
Molecular Formula | (S)nK2 |
Molecular Weight (g/mol) | NaN |
MDL Number | MFCD00148074 |
InChI Key | ZOCLAPYLSUCOGI-UHFFFAOYSA-N |
Synonym | potassium sulfane, potassium polysulfide, caswell no. 701, potassium sulfide k2 sx, epa pesticide chemical code 076703 |
PubChem CID | 5362529 |
SMILES | [K+].[K+].*-[S--]-* |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10774174
|
Thermo Scientific Acros
389592500 |
250 g | Plastic bottle |
42.51 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10711594
|
Thermo Scientific
389590010 |
1 kg | Plastic bottle |
107.54 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11974221
|
Thermo Scientific Acros
389590025 |
2.5 kg | Plastic bottle |
255.27 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
37199-66-9 | |
NaN | |
ZOCLAPYLSUCOGI-UHFFFAOYSA-N | |
5362529 |
(S)nK2 | |
MFCD00148074 | |
potassium sulfane, potassium polysulfide, caswell no. 701, potassium sulfide k2 sx, epa pesticide chemical code 076703 | |
[K+].[K+].*-[S--]-* |
Specifications
200.0°C to 250.0°C | |
(S)nK2 | |
MFCD00148074 | |
Solubility in water: 500g/l (20°C) | |
[K+].[K+].*-[S--]-* | |
NaN | |
Pure |
37199-66-9 | |
(K2S)n | |
potassium sulfane, potassium polysulfide, caswell no. 701, potassium sulfide k2 sx, epa pesticide chemical code 076703 | |
ZOCLAPYLSUCOGI-UHFFFAOYSA-N | |
potassium sulfane | |
5362529 | |
Potassium polysulfide |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
Contact with acids liberates toxic gas.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 253-390-1
- Potassium polysulfide
Signal Word
- Danger
Hazard Category
- ACUTE AQUATIC Acute 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H400-Very toxic to aquatic life.
- H314-Causes severe skin burns and eye damage.
- EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.